missing translation for 'onlineSavingsMsg'
Learn More

Ethanolamine, 99%, Thermo Scientific Chemicals

Product Code. 10103200
Click to view available options
Quantity:
250 mL
1 L
2.5 L
25 L

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-43-5
Molecular Formula C2H7NO
Molecular Weight (g/mol) 61.08
MDL Number MFCD00008183
InChI Key HZAXFHJVJLSVMW-UHFFFAOYSA-N
Synonym ethanolamine, monoethanolamine, aminoethanol, 2-hydroxyethylamine, colamine, glycinol, olamine, 2-amino-1-ethanol, ethanol, 2-amino, ethylolamine
PubChem CID 700
ChEBI CHEBI:16000
SMILES NCCO

Specifications

Melting Point 10.3°C
Density 1.0120g/mL
Boiling Point 170.0°C
Flash Point 92°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula H2NCH2CH2OH
Refractive Index 1.4530 to 1.4550
Quantity 2.5 L
Beilstein 04, 274
Fieser 01,357; 02,189; 04,222; 16,162
Merck Index 15, 3782
Specific Gravity 1.012
Solubility Information Solubility in water: soluble. Other solubilities: miscible with methanol and acetone, soluble in benzene, ether, ccl4 and n-heptane
IUPAC Name 2-aminoethanol
Viscosity 24 mPa.s (20°C)
Formula Weight 61.08
Vapor Pressure 0.48mmHg at 20°C
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Ethanolamine, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ethanolamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'