Learn More
Ethanolamine, 99%, Thermo Scientific Chemicals
26.40 € - 550.00 €
Chemical Identifiers
CAS | 141-43-5 |
---|---|
Molecular Formula | C2H7NO |
Molecular Weight (g/mol) | 61.08 |
MDL Number | MFCD00008183 |
InChI Key | HZAXFHJVJLSVMW-UHFFFAOYSA-N |
Synonym | ethanolamine, monoethanolamine, aminoethanol, 2-hydroxyethylamine, colamine, glycinol, olamine, 2-amino-1-ethanol, ethanol, 2-amino, ethylolamine |
PubChem CID | 700 |
ChEBI | CHEBI:16000 |
SMILES | NCCO |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10033213
|
Thermo Scientific Acros
149582500 |
250 mL |
26.40 €
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10133100
|
Thermo Scientific Acros
149580010 |
1 L |
49.70 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10103200
|
Thermo Scientific Acros
149580025 |
2.5 L |
94.90 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10552541
|
Thermo Scientific Acros
149580250 |
25 L |
550.00 €
25 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
141-43-5 | |
61.08 | |
HZAXFHJVJLSVMW-UHFFFAOYSA-N | |
700 | |
NCCO |
C2H7NO | |
MFCD00008183 | |
ethanolamine, monoethanolamine, aminoethanol, 2-hydroxyethylamine, colamine, glycinol, olamine, 2-amino-1-ethanol, ethanol, 2-amino, ethylolamine | |
CHEBI:16000 |
Specifications
141-43-5 | |
1.0120g/mL | |
92°C | |
98.5% min. (GC) | |
C2H7NO | |
1.4530 to 1.4550 | |
04, 274 | |
15, 3782 | |
ethanolamine, monoethanolamine, aminoethanol, 2-hydroxyethylamine, colamine, glycinol, olamine, 2-amino-1-ethanol, ethanol, 2-amino, ethylolamine | |
HZAXFHJVJLSVMW-UHFFFAOYSA-N | |
2-aminoethanol | |
700 | |
24 mPa.s (20°C) | |
0.48mmHg at 20°C | |
Liquid |
10.3°C | |
170.0°C | |
Authentic | |
Glass bottle | |
H2NCH2CH2OH | |
MFCD00008183 | |
01,357; 02,189; 04,222; 16,162 | |
1.012 | |
Solubility in water: soluble. Other solubilities: miscible with methanol and acetone, soluble in benzene, ether, ccl4 and n-heptane | |
NCCO | |
61.08 | |
CHEBI:16000 | |
61.08 | |
99% | |
Ethanolamine, 99% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes severe skin burns and eye damage.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED:
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 205-483-3
missing translation for 'rtecsNumber' : KJ5775000
missing translation for 'tsca' : TSCA
- Ethanolamine
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
- H412-Harmful to aquatic life with long lasting effects.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only