missing translation for 'onlineSavingsMsg'
Learn More

Ethanolamine hydrochloride, 98+%, Thermo Scientific Chemicals

Product Code. 11323466
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2002-24-6
Molecular Formula C2H8ClNO
Molecular Weight (g/mol) 97.542
MDL Number MFCD00012892
InChI Key PMUNIMVZCACZBB-UHFFFAOYSA-N
Synonym 2-aminoethanol hydrochloride, ethanolamine hydrochloride, ethanolamine hcl, mea hydrochloride, monoethanolamine hydrochloride, ethanolamine chloride, colamine hydrochloride, ethanol, 2-amino-, hydrochloride, ethanolamine-hcl, 2-hydroxyethylammonium chloride
PubChem CID 74819
IUPAC Name 2-aminoethanol;hydrochloride
SMILES C(CO)N.Cl

Specifications

Melting Point 83°C to 86°C
Quantity 500 g
Beilstein 3542892
Sensitivity Hygroscopic
Merck Index 14,3727
Formula Weight 97.55
Percent Purity ≥98%
Chemical Name or Material Ethanolamine hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.