missing translation for 'onlineSavingsMsg'
Learn More

Ether, Anhydrous, HPLC Grade, J.T. Baker™

Product Code. 15518554
Click to view available options
Quantity:
4 L
Packaging:
Narrow Mouth Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

High purity solvents for use in analytical and preparative separations

    For use in liquid chromatography (HPLC & UHPLC) & spectrophotometry, bulk pharmaceutical chemical

Chemical Identifiers

CAS 60-29-7
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
MDL Number MFCD00011646
InChI Key RTZKZFJDLAIYFH-UHFFFAOYSA-N
Synonym diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
PubChem CID 3283
ChEBI CHEBI:35702
IUPAC Name ethoxyethane
SMILES CCOCC

Specifications

Flash Point −45.00°C (113°F)
Packaging Narrow Mouth Amber Glass Bottle
Quantity 4 L
Specific Gravity 1L = 0.71kg
Formula Weight 74.12
Grade Meets USP requirements
Chemical Name or Material Ether, Anhydrous
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'