missing translation for 'onlineSavingsMsg'
Learn More
Ethyl 1,3-dithiane-2-carboxylate, 98+%, Thermo Scientific Chemicals Product Code.: 11360154

Ethyl 1,3-dithiane-2-carboxylate, 98+%, Thermo Scientific Chemicals

Product Code. 11360154
5 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11360154

Brand: Thermo Scientific Alfa Aesar L00663.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ethyl 1,3-dithiane-2-carboxylate is used in syn-selective aldol reactions. It undergoes asymmetric oxidation to give trans bis-sulfoxide. It is used to generate carbanon, which finds application in the preparation of alfa- keto esters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 1,3-dithiane-2-carboxylate is used in syn-selective aldol reactions. It undergoes asymmetric oxidation to give trans bis-sulfoxide. It is used to generate carbanon, which finds application in the preparation of alfa- keto esters.

Solubility
Slightly miscible with water.

Notes
Incompatible with acids, bases, oxidizing agents and reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20462-00-4
Molecular Formula C7H12O2S2
Molecular Weight (g/mol) 192.291
MDL Number MFCD00006657
InChI Key ANEDZEVDORCLPM-UHFFFAOYSA-N
Synonym 1,3-dithiane-2-carboxylic acid ethyl ester, carboethoxy-1,3-dithiane, 2-carboethoxy-1,3-dithiane, 1,3-dithiane-2-carboxylic acid, ethyl ester, 1,3 dithiane-2-carboxylic acid ethyl ester, pubchem22055, acmc-1cpb7, 2-carboethoxy 1,3-dithiane, 2-ethoxycarbonyl-1,3-dithiane, ethyl 1,3 dithiane carboxylate
PubChem CID 88552
IUPAC Name ethyl 1,3-dithiane-2-carboxylate
SMILES CCOC(=O)C1SCCCS1

Specifications

Melting Point 19°C to 21°C
Density 1.22
Boiling Point 75°C to 77°C (0.2 mmHg)
Flash Point 54°C (129°F)
Refractive Index 1.54
Quantity 5 g
UN Number UN1993
Beilstein 1424352
Solubility Information Slightly miscible with water.
Formula Weight 192.3
Percent Purity ≥98%
Chemical Name or Material Ethyl 1,3-dithiane-2-carboxylate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10250411 Certificate TypeCertificate of Analysis Date11/04/2024 Catalog NumberL00663.14,L00663.06
Lot Number10241072 Certificate TypeCertificate of Analysis Date02/06/2022 Catalog NumberL00663.14,L00663.06
Lot Number10234990 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL00663.14,L00663.06
Lot Number10143743 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL00663.14,L00663.06
Lot Number10153911 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL00663.14,L00663.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethyl 1,3-dithiane-2-carboxylate, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.