missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 1-(hydroxymethyl)cyclobutanecarboxylate, 97%

Product Code. 15459078
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15459078

Brand: Thermo Scientific Alfa Aesar H62566.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1195-81-9 | C8H14O3 | 158.197 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1195-81-9
Molecular Formula C8H14O3
Molecular Weight (g/mol) 158.197
MDL Number MFCD11707219
InChI Key PWMQFMMZBJUHID-UHFFFAOYSA-N
Synonym ethyl 1-hydroxymethyl cyclobutane-1-carboxylate, ethyl 1-hydroxymethylcyclobutanecarboxylate, ethyl 1-hydroxymethyl cyclobutanecarboxylate, cyclobutanecarboxylic acid, 1-hydroxymethyl-, ethyl ester, ethyl 1-hydroxymethyl-cyclobutanecarboxylate, 1-hydroxymethyl-cyclobutanecarboxylic acid ethyl ester
PubChem CID 15670506
IUPAC Name ethyl 1-(hydroxymethyl)cyclobutane-1-carboxylate
SMILES CCOC(=O)C1(CCC1)CO

Specifications

Quantity 5 g
Formula Weight 158.19
Percent Purity 97%
Chemical Name or Material Ethyl 1-(hydroxymethyl)cyclobutanecarboxylate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.