missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ethyl 2-(2-amino-1,3-thiazol-4-yl)acetate, Thermo Scientific™
Click to view available options
Quantity:
25 g
50 g
100 g
Specifications
Specifications
Assay Percent Range | 97% |
Quantity | 50 g |
Formula Weight | 186.23 |
Percent Purity | 97% |
Chemical Name or Material | Ethyl 2-(2-amino-1, 3-thiazol-4-yl)acetate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'