missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 2,4-dichloro-6-methylnicotinate, 97%, Thermo Scientific™
Click to view available options
Quantity:
5 g
10 g
25 g
Specifications
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Quantity | 5 g |
Formula Weight | 234.08 |
Percent Purity | 97% |
Chemical Name or Material | Ethyl 2, 4-dichloro-6-methylnicotinate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ethyl 2,4-dichloro-6-methylnicotinate, 97%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction