missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2,4-dioxo-4-(2-thienyl)butanoate, 90%, Thermo Scientific™

Product Code. 10059491
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 36983-36-5
Molecular Formula C10H10O4S
Molecular Weight (g/mol) 226.246
MDL Number MFCD00126316
InChI Key GCCFYXYKJZUHMI-UHFFFAOYSA-N
Synonym ethyl 2,4-dioxo-4-thiophen-2-yl butanoate, ethyl 2,4-dioxo-4-2-thienyl butanoate, 2,4-dioxo-4-thiophen-2-yl-butyric acid ethyl ester, 2-thiophenebutanoicacid, a,g-dioxo-, ethyl ester, 2,4-dioxo-4-2-thienyl butanoic acid ethyl ester, 2,4-dioxo-4-thiophen-2-ylbutyric acid ethyl ester
PubChem CID 2799459
IUPAC Name ethyl 2,4-dioxo-4-thiophen-2-ylbutanoate
SMILES CCOC(=O)C(=O)CC(=O)C1=CC=CS1

Specifications

CAS Min % 90.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 226.25
Percent Purity 90%
Chemical Name or Material Ethyl 2, 4-dioxo-4-(2-thienyl)butanoate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.