missing translation for 'onlineSavingsMsg'
Learn More
Ethyl 2-aminobenzoate, 99%, Thermo Scientific Chemicals Product Code.: 10226420

Ethyl 2-aminobenzoate, 99%, Thermo Scientific Chemicals

Product Code. 10226420
100 g, Glass bottle
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10226420

Brand: Thermo Scientific Acros 222231000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87-25-2
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.192
MDL Number MFCD00007711
InChI Key TWLLPUMZVVGILS-UHFFFAOYSA-N
Synonym ethyl anthranilate, ethyl o-aminobenzoate, anthranilic acid, ethyl ester, 2-aminobenzoic acid ethyl ester, 2-carboethoxyaniline, o-ethoxycarbonyl aniline, benzoic acid, 2-amino-, ethyl ester, ethylanthranilate, benzoic acid, o-amino-, ethyl ester, 2-ethoxycarbonyl anilineShow MoreShow Less
PubChem CID 6877
IUPAC Name ethyl 2-aminobenzoate
SMILES CCOC(=O)C1=CC=CC=C1N

Specifications

Color Yellow
Density 1.1170g/mL
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula H2NC6H4CO2C2H5
Refractive Index 1.5630 to 1.5650 (20°C, 589nm)
Quantity 100 g
Formula Weight 165.19
Percent Purity ≥98.5%
Physical Form Liquid
Chemical Name or Material Ethyl 2-aminobenzoate, 99%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0438961 Certificate TypeCertificate of Analysis Date10/02/2022 Catalog Number222235000
Lot NumberA0401066 Certificate TypeCertificate of Analysis Date08/04/2021 Catalog Number222235000,222231000
compliance-icons
Product Identifier
  • Ethyl 2-aminobenzoate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethyl 2-aminobenzoate, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.