missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-chloro-4-methyl-1,3-thiazole-5-carboxylate, 97%, Thermo Scientific™

Product Code. 10492674
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 7238-62-2
Molecular Formula C7H8ClNO2S
Molecular Weight (g/mol) 205.656
InChI Key VUARUZUFHDNJSY-UHFFFAOYSA-N
Synonym ethyl 2-chloro-4-methylthiazole-5-carboxylate, ethyl 2-chloro-4-methyl-5-thiazolecarboxylate, 5-thiazolecarboxylic acid, 2-chloro-4-methyl-, ethyl ester, 2-chloro-4-methyl-5-thiazolecarboxylic acid ethyl ester, 5-thiazolecarboxylicacid, 2-chloro-4-methyl-, ethyl ester, 2-chloro-4-methyl-thiazole-5-carboxylic acid ethyl ester
PubChem CID 594903
IUPAC Name ethyl 2-chloro-4-methyl-1,3-thiazole-5-carboxylate
SMILES CCOC(=O)C1=C(N=C(S1)Cl)C

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 205.66
Percent Purity 97%
Chemical Name or Material Ethyl 2-chloro-4-methyl-1, 3-thiazole-5-carboxylate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'