missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-chloro-4-methyl-1,3-thiazole-5-carboxylate, 97%, Thermo Scientific™

Product Code. 10516693
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 7238-62-2
Molecular Formula C7H8ClNO2S
Molecular Weight (g/mol) 205.656
InChI Key VUARUZUFHDNJSY-UHFFFAOYSA-N
Synonym ethyl 2-chloro-4-methylthiazole-5-carboxylate, ethyl 2-chloro-4-methyl-5-thiazolecarboxylate, 5-thiazolecarboxylic acid, 2-chloro-4-methyl-, ethyl ester, 2-chloro-4-methyl-5-thiazolecarboxylic acid ethyl ester, 5-thiazolecarboxylicacid, 2-chloro-4-methyl-, ethyl ester, 2-chloro-4-methyl-thiazole-5-carboxylic acid ethyl ester
PubChem CID 594903
IUPAC Name ethyl 2-chloro-4-methyl-1,3-thiazole-5-carboxylate
SMILES CCOC(=O)C1=C(N=C(S1)Cl)C

Specifications

Assay Percent Range 97%
Quantity 1 g
Formula Weight 205.66
Percent Purity 97%
Chemical Name or Material Ethyl 2-chloro-4-methyl-1, 3-thiazole-5-carboxylate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.