missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-cyclohexanoneacetate, 97%

Product Code. p-3757842 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 10566684

Brand: Thermo Scientific Acros 118320050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 24731-17-7 | C10H16O3 | 184.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24731-17-7
Molecular Formula C10H16O3
Molecular Weight (g/mol) 184.23
MDL Number MFCD00001638
InChI Key ZZWSNYNCRUZSPR-UHFFFAOYSA-N
Synonym ethyl 2-2-oxocyclohexyl acetate, ethyl 2-cyclohexanoneacetate, ethyl 2-oxocyclohexaneacetate, ethyl 2-oxocyclohexyl acetate, ethyl 2-cyclohexanone acetate, ethyl-2-cyclohexanone acetate, ethyl 2-oxocyclohexylacetate, cyclohexaneacetic acid, 2-oxo-, ethyl ester, ethyl 2-oxo-cyclohexyl-acetate
PubChem CID 91229
IUPAC Name ethyl 2-(2-oxocyclohexyl)acetate
SMILES CCOC(=O)CC1CCCCC1=O

Specifications

Color Colorless to Yellow
Density 1.0200g/mL
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (O=)C6H9CH2CO2C2H5
Refractive Index 1.4570 to 1.4590 (20°C, 589nm)
Quantity 5 g
Specific Gravity 1.02
Formula Weight 184.23
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Ethyl 2-cyclohexanoneacetate
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl 2-cyclohexanoneacetate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.