missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-hydroxyethyl sulfide, 98%, Thermo Scientific Chemicals

Product Code. 10687452
Click to view available options
Quantity:
5 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-77-0
Molecular Formula C4H10OS
Molecular Weight (g/mol) 106.18
MDL Number MFCD00002909
InChI Key LNRIEBFNWGMXKP-UHFFFAOYSA-N
Synonym 2-ethylthio ethanol, ethyl 2-hydroxyethyl sulfide, ethanol, 2-ethylthio, ethyl thioethanol, 2-hydroxyethyl ethyl sulfide, ethyl 2-hydroxyethyl thioether, beta-ethylthioethanol, 2-ethylsulfanyl ethanol, beta-hydroxydiethyl sulfide, 2-ethylthioethanol
PubChem CID 8075
IUPAC Name 2-ethylsulfanylethanol
SMILES CCSCCO

Specifications

Melting Point -100°C
Color Colorless to Yellow
Density 1.0200g/mL
Boiling Point 180°C to 184°C
Flash Point 93°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula C2H5SCH2CH2OH
Refractive Index 1.4859 to 1.4879
Quantity 5 g
Beilstein 01, II, 525
Merck Index 15, 3906
Specific Gravity 1.02
Formula Weight 106.18
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Ethyl 2-hydroxyethyl sulfide, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl 2-hydroxyethyl sulfide
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'