missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 2-(trifluoromethyl)-4-methyl-5-pyrimidine carboxylate, Thermo Scientific™
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
CAS | 306960-67-8 |
Packaging | Amber Glass Bottle |
Molecular Formula | C9H9F3N2O2 |
Quantity | 1 g |
Synonym | ethyl 4-methyl-2-trifluoromethyl pyrimidine-5-carboxylate, ethyl 2-trifluoromethyl-4-methyl-5-pyrimidine carboxylate, ethyl 2-trifluoromethyl-4-methylpyrimidine-5-carboxylate, ethyl-2-trifluoromethyl-4-methyl-5-pyrimidine carboxylate, ethyl4-methyl-2-trifluoromethyl pyrimidine-5-carboxylate, 5-pyrimidinecarboxylicacid, 4-methyl-2-trifluoromethyl-, ethyl ester |
InChI Key | LYEKBQSEKSEXLT-UHFFFAOYSA-N |
SMILES | CCOC(=O)C1=CN=C(N=C1C)C(F)(F)F |
IUPAC Name | ethyl 4-methyl-2-(trifluoromethyl)pyrimidine-5-carboxylate |
Molecular Weight (g/mol) | 234.178 |
PubChem CID | 10609709 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'