missing translation for 'onlineSavingsMsg'
Learn More
Ethyl 3-(2-furyl)propionate, 98%, Thermo Scientific Chemicals Product Code.: 11483008

Ethyl 3-(2-furyl)propionate, 98%, Thermo Scientific Chemicals

Product Code. 11483008
25 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11483008

Brand: Thermo Scientific Alfa Aesar A18732.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10031-90-0
Molecular Formula C9H12O3
Molecular Weight (g/mol) 168.192
MDL Number MFCD00036496
InChI Key OWIWZQQFSTZZIG-UHFFFAOYSA-N
Synonym ethyl 3-2-furyl propionate, ethyl 2-furanpropionate, 2-furanpropanoic acid, ethyl ester, ethyl 3-2-furyl propanoate, ethyl furfhydracrylate, ethyl furfurylacetate, ethyl 3-furan-2-yl propanoate, 2-furanpropionic acid, ethyl ester, ethyl 2-furanpropanoate, ethyl furan-2-propionate
PubChem CID 61450
IUPAC Name ethyl 3-(furan-2-yl)propanoate
SMILES CCOC(=O)CCC1=CC=CO1

Specifications

Quantity 25 g
Formula Weight 168.19
Percent Purity 98%
Chemical Name or Material Ethyl 3-(2-furyl)propionate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethyl 3-(2-furyl)propionate, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.