missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 3-[3,5-di(trifluoromethyl)phenyl]-3-oxopropanoate, 90%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
CAS Min % | 90.0 |
Assay Percent Range | 90% |
Packaging | Amber Glass Bottle |
Linear Formula | (CF3)2C6H3C(O)CH2CO2CH2CH3 |
Quantity | 1 g |
Formula Weight | 328.21 |
Percent Purity | 90% |
Chemical Name or Material | Ethyl 3-[3, 5-di(trifluoromethyl)phenyl]-3-oxopropanoate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'