missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3,5-dichloro-4-hydroxybenzoate, 99%, Thermo Scientific™

Product Code. 11484114
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Ethyl 3,5-dichloro-4-hydroxybenzoate is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Soluble in methanol.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17302-82-8
Molecular Formula C9H8Cl2O3
Molecular Weight (g/mol) 235.06
MDL Number MFCD00016420
InChI Key WMKNGSJJEMFQOT-UHFFFAOYSA-N
Synonym 3,5-dichloro-4-hydroxybenzoic acid ethyl ester, benzoic acid, 3,5-dichloro-4-hydroxy-, ethyl ester, 3,5-dichloro-4-hydroxy-benzoic acid ethyl ester, acmc-209e5q, 3-10-00-00363 beilstein handbook reference, wln: qr bg fg dvo2, ethyl 3,5-dichloro-4-hydroxy-benzoate, ethyl 3,5-bis chloranyl-4-oxidanyl-benzoate, benzoic acid,5-dichloro-4-hydroxy-, ethyl ester
PubChem CID 28460
IUPAC Name ethyl 3,5-dichloro-4-hydroxybenzoate
SMILES CCOC(=O)C1=CC(=C(C(=C1)Cl)O)Cl

Specifications

Melting Point 110°C to 112°C
Quantity 100 g
Beilstein 2649965
Solubility Information Soluble in methanol.
Formula Weight 235.07
Percent Purity 99%
Chemical Name or Material Ethyl 3,5-dichloro-4-hydroxybenzoate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'