missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3,5-dinitrobenzoate, 98%, Thermo Scientific™

Product Code. 11457184
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 618-71-3
Molecular Formula C9H8N2O6
Molecular Weight (g/mol) 240.17
MDL Number MFCD00007232
InChI Key IBQREHJPMPCXQA-UHFFFAOYSA-N
PubChem CID 69255
IUPAC Name ethyl 3,5-dinitrobenzoate
SMILES CCOC(=O)C1=CC(=CC(=C1)[N+]([O-])=O)[N+]([O-])=O

Specifications

Melting Point 91°C to 92°C
Quantity 50 g
Beilstein 1995473
Synonym benzoic acid, 3,5-dinitro-, ethyl ester, acmc-20al01, ethyl 3,5-dinitrobenzoate, 3,5-dinitrobenzoic acid ethyl ester, 3,5-dinitro-benzoic acid ethyl ester, benzoic acid,5-dinitro-, ethyl ester, benzoic acid,3,5-dinitro-, ethyl ester
Formula Weight 240.17
Percent Purity 98%
Chemical Name or Material Ethyl 3,5-dinitrobenzoate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.