missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-amino-1H-pyrazole-4-carboxylate, 99%, Thermo Scientific Chemicals

59.90 € - 899.00 €

Chemical Identifiers

CAS 6994-25-8
Molecular Formula C6H9N3O2
Molecular Weight (g/mol) 155.157
MDL Number MFCD00005238
InChI Key YPXGHKWOJXQLQU-UHFFFAOYSA-N
Synonym ethyl 3-amino-1h-pyrazole-4-carboxylate, ethyl 3-amino-4-pyrazolecarboxylate, 3-amino-4-carbethoxypyrazole, 5-amino-1h-pyrazole-4-carboxylic acid ethyl ester, 3-amino-1h-pyrazole-4-carboxylic acid ethyl ester, ethyl 3-aminopyrazole-4-carboxylate, 3-amino-4-ethoxycarbonylpyrazole, unii-9gy5ha1gaa, 3-amino-4-carbethoxypyrazol, 1h-pyrazole-4-carboxylic acid, 3-amino-, ethyl ester
PubChem CID 81472
IUPAC Name ethyl 5-amino-1H-pyrazole-4-carboxylate
SMILES CCOC(=O)C1=C(NN=C1)N
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11494233
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A10882.06
5 g
59.90 €
5g
Please to purchase this item. Need a web account? Register with us today!
11404243
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A10882.14
25 g
258.00 €
25g
Please to purchase this item. Need a web account? Register with us today!
11414243
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A10882.22
100 g
899.00 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

6994-25-8
155.157
YPXGHKWOJXQLQU-UHFFFAOYSA-N
81472
CCOC(=O)C1=C(NN=C1)N
C6H9N3O2
MFCD00005238
ethyl 3-amino-1h-pyrazole-4-carboxylate, ethyl 3-amino-4-pyrazolecarboxylate, 3-amino-4-carbethoxypyrazole, 5-amino-1h-pyrazole-4-carboxylic acid ethyl ester, 3-amino-1h-pyrazole-4-carboxylic acid ethyl ester, ethyl 3-aminopyrazole-4-carboxylate, 3-amino-4-ethoxycarbonylpyrazole, unii-9gy5ha1gaa, 3-amino-4-carbethoxypyrazol, 1h-pyrazole-4-carboxylic acid, 3-amino-, ethyl ester
ethyl 5-amino-1H-pyrazole-4-carboxylate

Specifications

6994-25-8
C6H9N3O2
4964
YPXGHKWOJXQLQU-UHFFFAOYSA-N
ethyl 5-amino-1H-pyrazole-4-carboxylate
81472
99%
104°C to 107°C
MFCD00005238
ethyl 3-amino-1h-pyrazole-4-carboxylate, ethyl 3-amino-4-pyrazolecarboxylate, 3-amino-4-carbethoxypyrazole, 5-amino-1h-pyrazole-4-carboxylic acid ethyl ester, 3-amino-1h-pyrazole-4-carboxylic acid ethyl ester, ethyl 3-aminopyrazole-4-carboxylate, 3-amino-4-ethoxycarbonylpyrazole, unii-9gy5ha1gaa, 3-amino-4-carbethoxypyrazol, 1h-pyrazole-4-carboxylic acid, 3-amino-, ethyl ester
CCOC(=O)C1=C(NN=C1)N
155.157
155.16
Ethyl 3-amino-1H-pyrazole-4-carboxylate
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 230-262-3

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only