Learn More
Ethyl 3-aminobenzoate, methanesulfonic acid salt, 98%, Thermo Scientific Chemicals
62.91 € - 737.67 €
Chemical Identifiers
CAS | 886-86-2 |
---|---|
Molecular Formula | C9H11NO2·CH4O3S |
Molecular Weight (g/mol) | 261.29 |
MDL Number | MFCD00013176 |
InChI Key | FQZJYWMRQDKBQN-UHFFFAOYSA-N |
Synonym | ethyl 3-aminobenzoate methanesulfonate, tricaine, tricaine methanesulfonate, finquel, metacaine, tricaine mesylate, 3-aminobenzoic acid ethyl ester methanesulfonate, ethyl m-aminobenzoate methanesulfonate, ethyl 3-aminobenzoate methanesulfonic acid salt, 3-aminobenzoic acid ethyl ester methanesulfonate salt |
PubChem CID | 261501 |
IUPAC Name | ethyl 3-aminobenzoate;methanesulfonic acid |
SMILES | CCOC(=O)C1=CC(=CC=C1)N.CS(=O)(=O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10743661
|
Thermo Scientific Acros
118000100 |
10 g | Glass bottle |
62.91 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10132960
|
Thermo Scientific Acros
118000500 |
50 g | Plastic bottle |
205.02 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10105493
|
Thermo Scientific Acros
118002500 |
250 g | Plastic bottle |
737.67 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
886-86-2 | |
261.29 | |
FQZJYWMRQDKBQN-UHFFFAOYSA-N | |
261501 | |
CCOC(=O)C1=CC(=CC=C1)N.CS(=O)(=O)O |
C9H11NO2·CH4O3S | |
MFCD00013176 | |
ethyl 3-aminobenzoate methanesulfonate, tricaine, tricaine methanesulfonate, finquel, metacaine, tricaine mesylate, 3-aminobenzoic acid ethyl ester methanesulfonate, ethyl m-aminobenzoate methanesulfonate, ethyl 3-aminobenzoate methanesulfonic acid salt, 3-aminobenzoic acid ethyl ester methanesulfonate salt | |
ethyl 3-aminobenzoate;methanesulfonic acid |
Specifications
886-86-2 | |
White | |
98% | |
H2NC6H4CO2C2H5·CH3SO3H | |
14, 389 | |
ethyl 3-aminobenzoate methanesulfonate, tricaine, tricaine methanesulfonate, finquel, metacaine, tricaine mesylate, 3-aminobenzoic acid ethyl ester methanesulfonate, ethyl m-aminobenzoate methanesulfonate, ethyl 3-aminobenzoate methanesulfonic acid salt, 3-aminobenzoic acid ethyl ester methanesulfonate salt | |
FQZJYWMRQDKBQN-UHFFFAOYSA-N | |
ethyl 3-aminobenzoate;methanesulfonic acid | |
261501 | |
98% | |
Ethyl 3-aminobenzoate, methanesulfonic acid salt |
146.0°C to 150.0°C | |
Authentic | |
C9H11NO2·CH4O3S | |
MFCD00013176 | |
15, 9786 | |
(5% in Water) Clear solution | |
CCOC(=O)C1=CC(=CC=C1)N.CS(=O)(=O)O | |
261.29 | |
261.29 | |
Fine Powder |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 212-956-8
- Ethyl 3-aminobenzoate, methanesulfonic acid salt
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.