missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-aminocrotonate, 98.5%

Product Code. 10416920 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 10416920

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7318-00-5 | C6H11NO2 | 129.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7318-00-5
Molecular Formula C6H11NO2
Molecular Weight (g/mol) 129.16
MDL Number MFCD02730138
InChI Key YPMPTULBFPFSEQ-PLNGDYQASA-N
Synonym z-ethyl 3-aminobut-2-enoate, ethyl 3-aminocrotonate, ethyl 3-aminobut-2-enoate, ethyl 2z-3-aminobut-2-enoate, 2-butenoic acid, 3-amino-, ethyl ester, 2z, ethyl z-3-aminobut-2-enoate, ethyl 3-amino-2-butenoate, .beta.-aminocrotonic acid ethyl ester, 2-butenoic acid, 3-amino-, ethyl ester, ethyl .beta.-aminocrotonate
PubChem CID 643756
IUPAC Name ethyl (Z)-3-aminobut-2-enoate
SMILES CCOC(=O)C=C(C)N

Specifications

Melting Point 33.0°C to 35.0°C
Color Yellow
Density 1.0220g/mL
Boiling Point 210.0°C to 215.0°C
Flash Point 97°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula CH3C(NH2)=CHCO2C2H5
Refractive Index 1.5000 to 1.5020
Quantity 500 g
Beilstein 03, 654
Specific Gravity 1.022
Formula Weight 129.16
Percent Purity 98.5%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material Ethyl 3-aminocrotonate, 98.5%
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl 3-aminocrotonate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.