missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-aminocrotonate, 98+%, Thermo Scientific Chemicals

Product Code. 11439873
Click to view available options
Quantity:
25 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Ethyl 3-aminocrotonate, is used as an intermediate in organic synthesis. It is also used as an active ingredient in pharmaceutical synthesis and feed auxiliary agent. It is also used as an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 3-aminocrotonate, is used as an intermediate in organic synthesis. It is also used as an active ingredient in pharmaceutical synthesis and feed auxiliary agent. It is also used as an active pharmaceutical intermediate.

Solubility
Soluble in water (26 g/L) (25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Incompatible to acids, bases, oxidizing agents, reducing agents. Air & Moisture Sensitive & Hygroscopic.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 626-34-6
Molecular Formula C6H11NO2
Molecular Weight (g/mol) 129.16
MDL Number MFCD00008073,MFCD02730138
InChI Key YPMPTULBFPFSEQ-UHFFFAOYSA-N
Synonym z-ethyl 3-aminobut-2-enoate, ethyl 3-aminocrotonate, ethyl 3-aminobut-2-enoate, ethyl 2z-3-aminobut-2-enoate, 2-butenoic acid, 3-amino-, ethyl ester, 2z, ethyl z-3-aminobut-2-enoate, ethyl 3-amino-2-butenoate, .beta.-aminocrotonic acid ethyl ester, 2-butenoic acid, 3-amino-, ethyl ester, ethyl .beta.-aminocrotonate
PubChem CID 643756
IUPAC Name ethyl (Z)-3-aminobut-2-enoate
SMILES CCOC(=O)C=C(C)N

Specifications

Melting Point 29°C to 35°C
Density 1.022
Boiling Point 210°C to 215°C
Flash Point 97°C (206°F)
Refractive Index 1.499
Quantity 500 g
UN Number UN3263
Beilstein 471284
Sensitivity Air and moisture sensitive; Hygroscopic
Solubility Information Soluble in water (26g/L) (25°C).
Formula Weight 129.16
Percent Purity ≥98%
Chemical Name or Material Ethyl 3-aminocrotonate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.