missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-bromopropionate, 99%, Thermo Scientific Chemicals

Product Code. 10731434
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 539-74-2
Molecular Formula C5H9BrO2
Molecular Weight (g/mol) 181.03
MDL Number MFCD00000251
InChI Key FQTIYMRSUOADDK-UHFFFAOYSA-N
Synonym ethyl 3-bromopropionate, propanoic acid, 3-bromo-, ethyl ester, 3-bromopropionic acid ethyl ester, ethyl beta-bromopropionate, unii-9b28g9s1jv, ccris 9051, propionic acid, 3-bromo-, ethyl ester, ethyl .beta.-bromopropionate, 3-bromo-propionic acid ethyl ester, ethyl-3-bromopropionate
PubChem CID 68320
IUPAC Name ethyl 3-bromopropanoate
SMILES CCOC(=O)CCBr

Specifications

Color Colorless to Yellow
Density 1.4190g/mL
Boiling Point 69°C to 71°C (10.0 mmHg)
Flash Point 70°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Linear Formula CH2BrCH2COOC2H5
Refractive Index 1.4510 to 1.453
Quantity 500 g
Beilstein 02, 256
Merck Index 15, 1444
Specific Gravity 1.419
Formula Weight 181.03
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Ethyl 3-bromopropionate, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl 3-bromopropionate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'