missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-cyano-3-phenylpyruvate, 97%, Thermo Scientific Chemicals

Product Code. 11441550
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6362-63-6
Molecular Formula C12H11NO3
Molecular Weight (g/mol) 217.22
MDL Number MFCD00019801
InChI Key YWNRATDDUSMBPR-UHFFFAOYNA-N
Synonym ethyl 3-cyano-3-phenylpyruvate, ethyl phenylcyanopyruvate, cyanophenylpyruvic acid ethyl ester, ethyl cyanophenylpyruvate, ethyl alpha-cyanophenylpyruvate, pyruvic acid, cyanophenyl-, ethyl ester, ethyl .alpha.-cyanophenylpyruvate, wln: ncr x1v1uno2, 4-10-00-03440 beilstein handbook reference, benzenepropanoic acid, .beta.-cyano-.alpha.-oxo-, ethyl ester
PubChem CID 22845
IUPAC Name ethyl 3-cyano-2-oxo-3-phenylpropanoate
SMILES CCOC(=O)C(=O)C(C#N)C1=CC=CC=C1

Specifications

Melting Point 125°C to 129°C
Quantity 5 g
Formula Weight 217.23
Percent Purity 97%
Chemical Name or Material Ethyl 3-cyano-3-phenylpyruvate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'