missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4,4-difluorocyclohexanecarboxylate, 97%, Thermo Scientific Chemicals

Product Code. 11369969
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 178312-47-5
Molecular Formula C9H14F2O2
Molecular Weight (g/mol) 192.206
MDL Number MFCD03788494
InChI Key HZZDWLBBNSDYQM-UHFFFAOYSA-N
PubChem CID 2779138
IUPAC Name ethyl 4,4-difluorocyclohexane-1-carboxylate
SMILES CCOC(=O)C1CCC(CC1)(F)F

Specifications

Boiling Point 84°C to 86°C (8 mmHg)
Quantity 250 mg
Synonym ethyl 4,4-difluorocyclohexanecarboxylate, cyclohexanecarboxylic acid, 4,4-difluoro-, ethyl ester, ethyl4,4-difluorocyclohexanecarboxylate, 4,4-difluoro-1-ethoxycarbonyl cyclohexane, 4,4-difluoro-cyclohexanecarboxylic acid ethyl ester, ethyl 4,4-difluorocyclohexane carboxylate, ethyl-4,4-difluorocyclohexane carboxylate, cyclohexanecarboxylic acid, 4,4-difluoro-, ethyl ester 9ci, acmc-209xsu, 4-carbethoxy-1,1-difluorocyclohexane
Formula Weight 192.21
Percent Purity 97%
Chemical Name or Material Ethyl 4,4-difluorocyclohexanecarboxylate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.