missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Ethyl 4-(5-chlorocarbonyl-2-furyl)benzoate

Product Code. 11305399
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 333435-04-4
Molecular Formula C14H11ClO4
Molecular Weight (g/mol) 278.688
MDL Number MFCD02258008
InChI Key OPOPFTSILPZGEL-UHFFFAOYSA-N
Synonym ethyl 4-5-chlorocarbonyl-2-furyl benzoate, 4-5-chlorocarbonyl-furan-2-yl-benzoic acid ethyl ester, ethyl 4-5-chlorocarbonyl furan-2-yl benzoate, ethyl 4-5-carbonochloridoyl furan-2-yl benzoate, acmc-20amvh, ethyl 4-5-carbonochloridoylfuran-2-yl benzoate, 4-5-chlorocarbonylfuran-2-yl benzoic acid ethyl ester, benzoic acid,4-5-chlorocarbonyl-2-furanyl-, ethyl ester
PubChem CID 3131522
IUPAC Name ethyl 4-(5-carbonochloridoylfuran-2-yl)benzoate
SMILES CCOC(=O)C1=CC=C(C=C1)C2=CC=C(O2)C(=O)Cl

Specifications

Quantity 1 g
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 278.69
Chemical Name or Material Ethyl 4-(5-chlorocarbonyl-2-furyl)benzoate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'