missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-bromobenzoate, 98+%, Thermo Scientific Chemicals

Product Code. 11440173
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Ethyl 4-bromobenzoate undergoes reduction with potassium diisobutyl-t-butoxyaluminum hydride (PDBBA) at 0°C to yield aldehydes. Reaction of ethyl 4-bromobenzoate and substituted benzyl chloride with zinc dust and a Pd catalyst is reported.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 4-bromobenzoate undergoes reduction with potassium diisobutyl-t-butoxyaluminum hydride (PDBBA) at 0°C to yield aldehydes. Reaction of ethyl 4-bromobenzoate and substituted benzyl chloride with zinc dust and a Pd catalyst is reported.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5798-75-4
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.07
MDL Number MFCD00016329
InChI Key XZIAFENWXIQIKR-UHFFFAOYSA-N
Synonym 4-bromobenzoic acid ethyl ester, ethyl p-bromobenzoate, benzoic acid, 4-bromo-, ethyl ester, ethyl4-bromobenzoate, ethyl-4-bromobenzoate, p-ethoxycarbonyl phenyl bromide, benzoic acid, p-bromo-, ethyl ester, 4-bromo-benzoic acid ethyl ester, 4-bromobenzoic acid, ethyl ester, ethyl-p-bromobenzoate
PubChem CID 22043
IUPAC Name ethyl 4-bromobenzoate
SMILES CCOC(=O)C1=CC=C(Br)C=C1

Specifications

Density 1.403
Boiling Point 262°C to 264°C
Flash Point >110°C (230°F)
Refractive Index 1.546
Quantity 10 g
Beilstein 1867129
Merck Index 14,1408
Solubility Information Slightly soluble in water.
Formula Weight 229.07
Percent Purity ≥98%
Chemical Name or Material Ethyl 4-bromobenzoate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'