missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-bromobutyrate, 97+%

Product Code. 10458150 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
Unit Size:
100mL
2.5 L
500mL
This item is not returnable. View return policy

Product Code. 10458150

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2969-81-5 | C6H11BrO2 | 195.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2969-81-5
Molecular Formula C6H11BrO2
Molecular Weight (g/mol) 195.06
InChI Key XBPOBCXHALHJFP-UHFFFAOYSA-N
Synonym ethyl 4-bromobutyrate, 4-bromobutyric acid ethyl ester, ethyl-4-bromobutyrate, butanoic acid, 4-bromo-, ethyl ester, ethyl-4-bromotbutyrate, 4-bromo-n-butyric acid ethyl ester, brch2ch2ch2c o oc2h5, 4-bromo-butyric acid ethyl ester, 4-bromo-butanoic acid ethyl ester, gamma-bromobutyric acid ethyl ester
PubChem CID 76300
IUPAC Name ethyl 4-bromobutanoate
SMILES CCOC(=O)CCCBr

Specifications

Color Colorless to Yellow
Density 1.3600g/mL
Boiling Point 80.0°C to 82.0°C (10.0 mmHg)
Flash Point 90°C
Infrared Spectrum Authentic
Assay Percent Range 97% min. (GC)
Packaging Glass bottle
Linear Formula Br(CH2)3CO2C2H5
Refractive Index 1.4549 to 1.4569
Quantity 100 mL
Beilstein 02,283
Specific Gravity 1.36
Solubility Information Solubility in water: insoluble.
Formula Weight 195.06
Percent Purity 97+%
Physical Form Liquid
Chemical Name or Material Ethyl 4-bromobutyrate
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl 4-bromobutyrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.