missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-chloro-6-methoxyquinoline-3-carboxylate, 97%, Thermo Scientific™

70.50 € - 235.00 €

Chemical Identifiers

CAS 22931-71-1
Molecular Formula C13H12ClNO3
Molecular Weight (g/mol) 265.693
MDL Number MFCD00173417
InChI Key QURGQUFEJWWDRF-UHFFFAOYSA-N
Synonym gnf-pf-3551, 4-chloro-6-methoxy-quinoline-3-carboxylic acid ethyl ester, 4-chloro-6-methoxyquinoline-3-carboxylic acid ethyl ester, ethyl 4-chloro-6-methoxy-3-quinolinecarboxylate, acmc-20a6z6, 3-carbethoxy-4-chloro-6-methoxyquinoline, 4-chloro-3-ethoxycarbonyl-6-methoxyquinoline, 6-methoxy-4-chloro-3-ethoxycarbonylquinoline, ethyl 4chloro-6methoxyquinoline-3-carboxylate, ethyl 4-chloro-6-methoxyquinolin-3-carboxylate
PubChem CID 304558
IUPAC Name ethyl 4-chloro-6-methoxyquinoline-3-carboxylate
SMILES CCOC(=O)C1=CN=C2C=CC(=CC2=C1Cl)OC
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11375599
View Documents
Thermo Scientific Alfa Aesar
H50515.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11385599
View Documents
Thermo Scientific Alfa Aesar
H50515.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Ethyl 4-chloro-6-methoxyquinoline-3-carboxylate is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Notes
Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.
Specifications

Chemical Identifiers

22931-71-1
265.693
QURGQUFEJWWDRF-UHFFFAOYSA-N
304558
CCOC(=O)C1=CN=C2C=CC(=CC2=C1Cl)OC
C13H12ClNO3
MFCD00173417
gnf-pf-3551, 4-chloro-6-methoxy-quinoline-3-carboxylic acid ethyl ester, 4-chloro-6-methoxyquinoline-3-carboxylic acid ethyl ester, ethyl 4-chloro-6-methoxy-3-quinolinecarboxylate, acmc-20a6z6, 3-carbethoxy-4-chloro-6-methoxyquinoline, 4-chloro-3-ethoxycarbonyl-6-methoxyquinoline, 6-methoxy-4-chloro-3-ethoxycarbonylquinoline, ethyl 4chloro-6methoxyquinoline-3-carboxylate, ethyl 4-chloro-6-methoxyquinolin-3-carboxylate
ethyl 4-chloro-6-methoxyquinoline-3-carboxylate

Specifications

22931-71-1
C13H12ClNO3
gnf-pf-3551, 4-chloro-6-methoxy-quinoline-3-carboxylic acid ethyl ester, 4-chloro-6-methoxyquinoline-3-carboxylic acid ethyl ester, ethyl 4-chloro-6-methoxy-3-quinolinecarboxylate, acmc-20a6z6, 3-carbethoxy-4-chloro-6-methoxyquinoline, 4-chloro-3-ethoxycarbonyl-6-methoxyquinoline, 6-methoxy-4-chloro-3-ethoxycarbonylquinoline, ethyl 4chloro-6methoxyquinoline-3-carboxylate, ethyl 4-chloro-6-methoxyquinolin-3-carboxylate
CCOC(=O)C1=CN=C2C=CC(=CC2=C1Cl)OC
265.693
265.7
Ethyl 4-chloro-6-methoxyquinoline-3-carboxylate
88°C to 90°C
MFCD00173417
QURGQUFEJWWDRF-UHFFFAOYSA-N
ethyl 4-chloro-6-methoxyquinoline-3-carboxylate
304558
97%
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications