missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-ethoxybenzoate, 98%, Thermo Scientific Chemicals

Product Code. 11447247
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Not miscible or difficult to mix with water.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23676-09-7
Molecular Formula C11H14O3
Molecular Weight (g/mol) 194.23
MDL Number MFCD00009116
InChI Key HRAQMGWTPNOILP-UHFFFAOYSA-N
Synonym benzoic acid, 4-ethoxy-, ethyl ester, 4-ethoxybenzoic acid ethyl ester, 4-ethoxy ethylbenzoate, benzoic acid, p-ethoxy-, ethyl ester, 4-ethoxyethylbenzoate, peeb, ethyl p-ethoxybenzoate, ethyl-4-ethoxybenzoate, p-ethoxy ethyl benzoate, ethyl-p-ethoxy benzoate
PubChem CID 90232
IUPAC Name ethyl 4-ethoxybenzoate
SMILES CCOC1=CC=C(C=C1)C(=O)OCC

Specifications

Melting Point 9°C
Density 1.071
Boiling Point 145°C to 150°C (13 mmHg)
Flash Point 137°C (278°F)
Refractive Index 1.518
Quantity 25 g
Beilstein 2692236
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 194.23
Percent Purity 98%
Chemical Name or Material Ethyl 4-ethoxybenzoate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.