missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-methyl-1,2,3-thiadiazole-5-carboxylate, 98%, Thermo Scientific Chemicals

Product Code. 11415413
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18212-20-9
Molecular Formula C6H8N2O2S
Molecular Weight (g/mol) 172.20
MDL Number MFCD00052549
InChI Key AHPXTXGCMLOXGA-UHFFFAOYSA-N
Synonym ethyl 4-methyl-1,2,3-thiadiazole-5-carboxylate, 4-methyl-1,2,3-thiadiazole-5-carboxylic acid ethylester, 1,2,3-thiadiazole-5-carboxylic acid, 4-methyl-, ethyl ester, acmc-209xfp, ethylmethylthiadiazolecarboxylate, 5-ethoxycarbonyl-4-methyl-1,2,3-thiadiazole, 4-methyl-5-thiadiazolecarboxylic acid ethyl ester, 4-methyl-1,2,3 thiadiazole-5-carboxylic acid ethyl ester, 4-methyl-1,2,3-thiadiazole-5-carboxylic acid ethyl ester
PubChem CID 2758878
IUPAC Name ethyl 4-methylthiadiazole-5-carboxylate
SMILES CCOC(=O)C1=C(C)N=NS1

Specifications

Density 1.265
Boiling Point 117°C to 118°C (15 mmHg)
Refractive Index 1.504
Quantity 1 g
Beilstein 609140
Formula Weight 172.21
Percent Purity 98%
Chemical Name or Material Ethyl 4-methyl-1,2,3-thiadiazole-5-carboxylate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'