missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 5-bromovalerate, 98%, Thermo Scientific Chemicals

56.18 € - 651.24 €

Chemical Identifiers

CAS 14660-52-7
Molecular Formula C7H13BrO2
Molecular Weight (g/mol) 209.08
MDL Number MFCD00000266
InChI Key AFRWBGJRWRHQOV-UHFFFAOYSA-N
PubChem CID 84580
IUPAC Name ethyl 5-bromopentanoate
SMILES CCOC(=O)CCCCBr
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11493954
View Documents
Thermo Scientific Alfa Aesar
A11952.14
25 g
56.18 €
25g
Please to purchase this item. Need a web account? Register with us today!
11403964
View Documents
Thermo Scientific Alfa Aesar
A11952.22
100 g
173.87 €
100g
Please to purchase this item. Need a web account? Register with us today!
11413964
View Documents
Thermo Scientific Alfa Aesar
A11952.36
500 g
651.24 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Ethyl 5-bromovalerate is employed in the preparation of S(γ-Carboxypropyl)-DL-homocysteine and S(δ-Carboxybutyl)-DL-homocysteine. It is also used in the preparation of ethyl 5-azidovalerate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 5-bromovalerate is employed in the preparation of S(γ-Carboxypropyl)-DL-homocysteine and S(δ-Carboxybutyl)-DL-homocysteine. It is also used in the preparation of ethyl 5-azidovalerate.

Solubility
Not miscible or difficult to mix in water.

Notes
Light Sensitive. Keep away from oxidizing agents. Store in dark.
Specifications

Chemical Identifiers

14660-52-7
209.08
AFRWBGJRWRHQOV-UHFFFAOYSA-N
ethyl 5-bromopentanoate
C7H13BrO2
MFCD00000266
84580
CCOC(=O)CCCCBr

Specifications

14660-52-7
227°C
C7H13BrO2
1.458
1752023
ethyl 5-bromovalerate, 5-bromopentanoic acid ethyl ester, pentanoic acid, 5-bromo-, ethyl ester, 5-bromovaleric acid ethyl ester, ethyl omega-bromovalerate, ethyl5-bromovalerate, ethyl-5-bromovalerate, pubchem3955, ethyl-5-bromo-valerate, ethyl-5-bromopentanoate
AFRWBGJRWRHQOV-UHFFFAOYSA-N
ethyl 5-bromopentanoate
84580
98%
1.306
103°C (217°F)
Br(CH2)4CO2CH2CH3
MFCD00000266
Light sensitive
Not miscible or difficult to mix in water.
CCOC(=O)CCCCBr
209.08
209.09
Ethyl 5-bromovalerate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 238-705-2

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

Product Identifier
  • Ethyl 5-bromovalerate

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Supplemental information
  • MIXTURE LIST-Contains : Ethyl 5-bromovalerate

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.