missing translation for 'onlineSavingsMsg'
Learn More
Ethyl 5-(chlorosulfonyl)-3-furoate, 95%, Thermo Scientific™ Product Code.: 10285504

Ethyl 5-(chlorosulfonyl)-3-furoate, 95%, Thermo Scientific™

Product Code. 10285504
5 g, Amber Glass Bottle
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 10285504

Brand: Thermo Scientific Maybridge CC73303DE

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 256373-91-8
Molecular Formula C7H7ClO5S
Molecular Weight (g/mol) 238.638
MDL Number MFCD11109333
InChI Key LEKOVVUPHAKDKN-UHFFFAOYSA-N
Synonym ethyl 5-chlorosulfonyl furan-3-carboxylate, ethyl 5-chlorosulfonyl-3-furoate, 3-furancarboxylic acid,5-chlorosulfonyl-, ethyl ester, 5-chlorosulfonyl-furan-3-carboxylic acid ethyl ester, 2-chlorosulfonyl furan-4-carboxylic acid ethyl ester, 3-furancarboxylic acid, 5-chlorosulfonyl-, ethyl ester
PubChem CID 10130959
IUPAC Name ethyl 5-chlorosulfonylfuran-3-carboxylate
SMILES CCOC(=O)C1=COC(=C1)S(=O)(=O)Cl

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 238.65
Percent Purity 95%
Chemical Name or Material Ethyl 5-(chlorosulfonyl)-3-furoate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethyl 5-(chlorosulfonyl)-3-furoate, 95%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.