missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 5-fluoroindole-2-carboxylate, 98%, Thermo Scientific Chemicals

Product Code. 11378885
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 348-36-7
Molecular Formula C11H10FNO2
Molecular Weight (g/mol) 207.20
MDL Number MFCD00152076
InChI Key VIKOQTQMWBKMNA-UHFFFAOYSA-N
Synonym ethyl 5-fluoroindole-2-carboxylate, 5-fluoroindole-2-carboxylic acid ethyl ester, 2-carbethoxy-5-fluoroindole, 1h-indole-2-carboxylic acid, 5-fluoro-, ethyl ester, 5-fluoro-1h-indole-2-carboxylic acid ethyl ester, ethyl5-fluoro-indole-2-carboxylate, pubchem7276, maybridge1_006190, acmc-1aex1, ksc573a9h
PubChem CID 2727899
IUPAC Name ethyl 5-fluoro-1H-indole-2-carboxylate
SMILES CCOC(=O)C1=CC2=CC(F)=CC=C2N1

Specifications

Melting Point 147°C to 152°C
Quantity 1 g
Beilstein 170254
Formula Weight 207.21
Percent Purity 98%
Chemical Name or Material Ethyl 5-fluoroindole-2-carboxylate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'