missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 7-bromoheptanoate, 97%, Thermo Scientific Chemicals

Product Code. 15493807
Click to view available options
Quantity:
5 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 29823-18-5
Density 1.217
Boiling Point 112°C (5 mmHg)
Flash Point >110°C (230°F)
Molecular Formula C9H17BrO2
MDL Number MFCD00037103
Quantity 5 mL
Synonym 7-bromoheptanoic acid ethyl ester, ethyl-7-bromoheptanoate, heptanoic acid, 7-bromo-, ethyl ester, ethyl7-bromoheptanoate, br ch2 6cooet, ethyl 7-bromo-heptanoate, ethyl 7-bromanylheptanoate, acmc-209hb9, ethyl 7-bromoheptanoate, oobfndgmagsnka-uhfffaoysa
InChI Key OOBFNDGMAGSNKA-UHFFFAOYSA-N
SMILES CCOC(=O)CCCCCCBr
IUPAC Name ethyl 7-bromoheptanoate
Molecular Weight (g/mol) 237.14
PubChem CID 122490
Formula Weight 237.14
Percent Purity 97%
Chemical Name or Material Ethyl 7-bromoheptanoate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'