missing translation for 'onlineSavingsMsg'
Learn More
Ethyl acetate, 99.8%, for biochemistry, AcroSeal™, Thermo Scientific Chemicals Product Code.: 12652690

Ethyl acetate, 99.8%, for biochemistry, AcroSeal™, Thermo Scientific Chemicals

Product Code. 12652690
2.5 L, AcroSeal™ Glass Bottle
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
AcroSeal™ Glass Bottle
This item is not returnable. View return policy

Product Code. 12652690

Brand: Thermo Scientific Acros 364240025

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-78-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
InChI Key XEKOWRVHYACXOJ-UHFFFAOYSA-N
Synonym ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic esterShow MoreShow Less
PubChem CID 8857
ChEBI CHEBI:27750
IUPAC Name ethyl acetate
SMILES CCOC(=O)C

Specifications

Melting Point -83.5°C
Absorbance 0.03 max. at 280 nm, 0.05 max. at 265 nm, 0.1 max. at 263 nm, 0.3 max. at 260 nm, 1 max. at 255 nm
Density 0.9020g/mL
Boiling Point 75.0°C to 78.0°C
Quantity 2.5 L
Flash Point −4°C
Packaging AcroSeal™ Glass Bottle
Refractive Index 1.3710 to 1.3740
Assay Percent Range 0.998
Specific Gravity 0.902
Linear Formula CH3CO2C2H5
Viscosity 0.45 mPa.s (20°C)
Solubility Information Solubility in water: 80g/L (20°C). Other solubilities: soluble in benzene, acetone, chloroform, alcohol,, ether
Formula Weight 88.11
Percent Purity 99.8%
Grade Biochemical
Chemical Name or Material Ethyl acetate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0460652 Certificate TypeCertificate of Analysis Date15/03/2024 Catalog Number364240010,364240025
Lot NumberA0453099 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number364240010,364240025
Lot NumberA0450976 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number364240010,364240025
Lot NumberA0396887 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number364240010,364240025
Lot NumberA0412074 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number364240010,364240025
compliance-icons compliance-icons
Product Identifier
  • Ethyl acetate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • H225-Highly flammable liquid and vapour.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethyl acetate, 99.8%, for biochemistry, AcroSeal™, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.