Learn More
Ethyl acetate, 99%, Thermo Scientific Chemicals
Description
Ethyl acetate is widely used as a solvent in cleaning of the circuit board. It is most suited for HPLC instruments and spectrophotometers including the UV-visible spectrophotometer, the IR spectrometer and the mass spectrometer. In addition, it can also be used as a solvent in column chromatography and solvent extraction. It is also used in glues, nail polish removers, decaffeinating tea and coffee and in cigarettes. It is present in confectioneries, perfumes and fruits.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Ethyl acetate is widely used as a solvent in cleaning of the circuit board. It is most suited for HPLC instruments and spectrophotometers including the UV-visible spectrophotometer, the IR spectrometer and the mass spectrometer. In addition, it can also be used as a solvent in column chromatography and solvent extraction. It is also used in glues, nail polish removers, decaffeinating tea and coffee and in cigarettes. It is present in confectioneries, perfumes and fruits.
Solubility
Miscible with ethanol, acetone, diethyl ether and benzene.
Notes
It is flammable and may be moisture sensitive.

Specifications
Specifications
Melting Point | -84°C |
Density | 0.902 |
Boiling Point | 76°C to 77°C |
Quantity | 250 mL |
Flash Point | −3°C (26°F) |
Odor | Sweet |
Refractive Index | 1.372 |
Linear Formula | CH3CO2CH2CH3 |
UN Number | UN1173 |
Beilstein | 506104 |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2409982 | Certificate of Analysis | 15/11/2024 | L10925.0F,L10925.AU,L10925.AK |
2401642 | Certificate of Analysis | 04/10/2024 | L10925.0F,L10925.AU,L10925.AK |
2482209 | Certificate of Analysis | 30/08/2024 | L10925.0F,L10925.AU,L10925.AK |
2404441 | Certificate of Analysis | 09/08/2024 | L10925.0F,L10925.AU,L10925.AK |
2484809 | Certificate of Analysis | 22/02/2024 | L10925.0F,L10925.AU,L10925.AK |
Safety and Handling


- Ethyl acetate
- Danger
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- EUH066-Repeated exposure may cause skin dryness or cracking.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contains : Ethyl acetate
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.