Learn More
Ethyl acetate, ACS, 99.5+%, Thermo Scientific Chemicals
Description
Ethyl acetate is widely used as a solvent in cleaning of the circuit board. It is most suited for HPLC instruments and spectrophotometers including the UV-visible spectrophotometer, the IR spectrometer and the mass spectrometer. In addition, it can also be used as a solvent in column chromatography and solvent extraction. It is also used in glues, nail polish removers, decaffeinating tea and coffee and in cigarettes. It is present in confectioneries, perfumes and fruits.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | -84°C |
Color | Colorless |
Density | 0.902 g/mL |
Boiling Point | 77°C |
Quantity | 1 L |
Flash Point | -3°C (26°F) |
Odor | Sweet |
Physical Form | Liquid |
Refractive Index | 1.372 |
Linear Formula | CH3CO2CH2CH3 |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
N28L005 | Certificate of Analysis | 03/03/2025 | 031344.M1,031344.AP,031344.K7,031344.K2,031344.M4 |
N14L031 | Certificate of Analysis | 17/02/2025 | 031344.M1,031344.AP,031344.K7,031344.K2,031344.M4 |
M31L047 | Certificate of Analysis | 03/02/2025 | 031344.M1,031344.AP,031344.K7,031344.K2,031344.M4 |
Y25K007 | Certificate of Analysis | 26/11/2024 | 031344.M1,031344.AP,031344.K7,031344.K2,031344.M4 |
X04K034 | Certificate of Analysis | 15/10/2024 | 031344.M1,031344.AP,031344.K7,031344.K2,031344.M4 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.