missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Acetate, Certified AR for Analysis, Fisher Chemical™
C4H8O2, CAS Number-141-78-6, acetic acid ethyl ester, acetic acid, ethyl ester, acetic ester, acetic ether, acetidin, acetoxyethane, ethylacetate, ethyl acetic ester, ethyl ethanoate, vinegar naphtha, 2.5L, 75 deg.C, CHEBI:27750, Colorless, 88.11g/mol, XEKOWRVHYACXOJ-UHFFFAOYSA-N, ethyl acetate
51.70 € - 510.00 €
Chemical Identifiers
CAS | 141-78-6 |
---|---|
Molecular Formula | C4H8O2 |
MDL Number | 9171 |
InChI Key | XEKOWRVHYACXOJ-UHFFFAOYSA-N |
PubChem CID | 8857 |
ChEBI | CHEBI:27750 |
IUPAC Name | ethyl acetate |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10697212
|
Fisher Chemical
E/0900/15 |
1 L | Amber Glass Bottle |
51.70 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10386320
|
Fisher Chemical
E/0900/17 |
2.5 L | Amber Glass Bottle |
125.00 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10388003
|
Fisher Chemical
E/0900/PC17 |
2.5 L | HDPE Bottle |
107.00 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10429750
|
Fisher Chemical
E/0900/PB17 |
2.5 L | Plastic-coated Glass Bottle |
117.00 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10182392
|
Fisher Chemical
E/0900/21 |
5 L | Metal Can |
169.00 €
5 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12058598
|
Fisher Chemical
E/0900/FP21 |
5 L | Plastic Container |
167.00 €
5 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10396320
|
Fisher Chemical
E/0900/25 |
25 L | Metal Can |
510.00 €
25 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
C4H8O2, CAS Number-141-78-6, acetic acid ethyl ester, acetic acid, ethyl ester, acetic ester, acetic ether, acetidin, acetoxyethane, ethylacetate, ethyl acetic ester, ethyl ethanoate, vinegar naphtha, 2.5L, 75 deg.C, CHEBI:27750, Colorless, 88.11g/mol, XEKOWRVHYACXOJ-UHFFFAOYSA-N, ethyl acetate
Chemical Identifiers
141-78-6 | |
9171 | |
8857 | |
ethyl acetate |
C4H8O2 | |
XEKOWRVHYACXOJ-UHFFFAOYSA-N | |
CHEBI:27750 |
Specifications
-83.5°C | |
Colorless | |
Liquid | |
9171 | |
0.45cp at 20°C | |
CCOC(=O)C | |
88.106 | |
CHEBI:27750 |
141-78-6 | |
75°C | |
C4H8O2 | |
103 mbar at 20°C | |
XEKOWRVHYACXOJ-UHFFFAOYSA-N | |
ethyl acetate | |
8857 | |
88.11g/mol |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Ethyl acetate
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : Ethyl acetate