missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Acetate Certified AR, Fisher Chemical™

Ethyl acetate Certified AR 200L Conforms to ACS specifications

Brand:  Fisher Chemical E/0900/27

849.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.



Product Code. 10238583

  • 850.00 € / 200 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Ethyl acetate Certified AR 200L Conforms to ACS specifications

  • Ethyl acetate Certified AR 200L Conforms to ACS specifications Plastic Bottle Cas No. 141-78-6
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Ethyl Acetate
141-78-6
200 L
C4H8O2
CCOC(C)=O
88.11
-84°C (literature)
77°C
Plastic
XEKOWRVHYACXOJ-UHFFFAOYSA-N
ethyl acetate
Product Suggestions

Product Suggestions

Videos
Safety and Handling
Show More
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2534843 Certificate TypeCertificate of Analysis Date14/04/2025 Catalog NumberE/0900/17,E/0900/17Z,E/0900/PC17,E/0900/27,E/0900/15,E/0900/PB17,E/0900/21Z,E/0900/PB17X,E/0900/25
Lot Number2533486 Certificate TypeCertificate of Analysis Date04/04/2025 Catalog NumberE/0900/17,E/0900/17Z,E/0900/PC17,E/0900/27,E/0900/15,E/0900/PB17,E/0900/21Z,E/0900/PB17X,E/0900/25
Lot Number2529102 Certificate TypeCertificate of Analysis Date28/02/2025 Catalog NumberE/0900/17,E/0900/17Z,E/0900/PC17,E/0900/27,E/0900/15,E/0900/PB17,E/0900/21Z,E/0900/PB17X,E/0900/25
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethyl Acetate Certified AR, Fisher Chemical™ > Quantity: 200L, Packaging: Plastic

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.