missing translation for 'onlineSavingsMsg'
Learn More

Ethyl acetate, Environmental Grade, 99.5+%, Thermo Scientific Chemicals

Product Code. 11305317
1 L
Click to view available options
Quantity:
1 L
4 x 1 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Pesticide residue, environmental, and GC analysis

Ethyl acetate is used primarily as a solvent and diluent. It is commonly used to clean the circuit boards, in some nail varnish removers and also in residue, environmental and GC analysis. It is used chiefly as a scent in the manufacture of perfumes. It is also used in flavorings, glues, in decaffeinating tea and coffee, and in cigarettes. The other minor applications of ethyl acetate include its applications in the textiles, dyestuffs, explosives, photographic films and plates, artificial leather.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-78-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
MDL Number MFCD00009171
InChI Key XEKOWRVHYACXOJ-UHFFFAOYSA-N
Synonym ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
PubChem CID 8857
ChEBI CHEBI:27750
IUPAC Name ethyl acetate
SMILES CCOC(=O)C

Specifications

Melting Point -84°C
Color Colorless
Density 0.902 g/mL
Boiling Point 77°C
Quantity 1 L
Flash Point -3°C (26°F)
Odor Sweet
Physical Form Liquid
Refractive Index 1.372
Linear Formula CH3CO2CH2CH3
UN Number UN1173
Beilstein 506104
Merck Index 14,3757
Solubility Information Miscible with ethanol,acetone,diethyl ether and benzene.
Formula Weight 88.11
Percent Purity ≥99.5%
Grade Environmental
Chemical Name or Material Ethyl acetate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethyl acetate, Environmental Grade, 99.5+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.