missing translation for 'onlineSavingsMsg'
Learn More
Ethyl acetate, HPLC Grade, 99.5+%, Thermo Scientific Chemicals Product Code.: 11338167

Ethyl acetate, HPLC Grade, 99.5+%, Thermo Scientific Chemicals

Product Code. 11338167
1 L
Click to view available options
Quantity:
1 L
4 x 1 L
This item is not returnable. View return policy

Product Code. 11338167

Brand: Thermo Scientific Alfa Aesar 022912.K2

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

Ethyl acetate in its highly pure form is used as a solvent for spectrophotometric applications. It is most suited for HPLC instruments and spectrophotometers including UV-visible spectrophotometer, IR spectrometer and mass spectrometer. In addition, it can also be used as a solvent in column chromatography and solvent extraction. Also, it is used in glues, nail polish removers, decaffeinating tea and coffee, and cigarettes. It is present in confectioneries, perfumes, and fruits.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-78-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
MDL Number MFCD00009171
InChI Key XEKOWRVHYACXOJ-UHFFFAOYSA-N
Synonym ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
PubChem CID 8857
ChEBI CHEBI:27750
IUPAC Name ethyl acetate
SMILES CCOC(=O)C

Specifications

Melting Point °C to 84°C
Color Colorless
Appearance Colorless liquid
Density 0.902 g/cm3
Boiling Point 77°C
Quantity 1 L
Flash Point -3°C (26°F)
Ignition Point 460°C (860°F)
Odor Fruit-like
Physical Form Liquid
Refractive Index 1.372
Assay Percent Range >99.5%
Beilstein 506104
Solubility Information Miscible with ethanol,acetone,diethyl ether and benzene.
Formula Weight 88.11
Grade HPLC
Chemical Name or Material Ethyl acetate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberM30L714 Certificate TypeCertificate of Analysis Date05/03/2025 Catalog Number022912.K7,022912.M1,022912.K2
Lot NumberIY732463 Certificate TypeCertificate of Analysis Date13/09/2024 Catalog Number022912.K7,022912.M1,022912.K2
Lot NumberQ05K781 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number022912.K7,022912.M1,022912.K2
Lot NumberBS732403 Certificate TypeCertificate of Analysis Date20/03/2024 Catalog Number022912.K7,022912.M1,022912.K2
Lot NumberMH740442 Certificate TypeCertificate of Analysis Date10/08/2023 Catalog Number022912.K7,022912.M1,022912.K2
compliance-icons compliance-icons
Product Identifier
  • Ethyl acetate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P242-Use only non-sparking tools.
  • P243-Take precautionary measures against static discharge.
  • P264-Wash thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P370+P378-In case of fire: Use dry sand, dry chemical, or alcohol-resistant foam to extinguish.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : ethyl acetate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethyl acetate, HPLC Grade, 99.5+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.