Learn More
Ethyl acetoacetate, 99+%, Thermo Scientific Chemicals
24.82 € - 274.37 €
Chemical Identifiers
CAS | 141-97-9 |
---|---|
Molecular Formula | C6H10O3 |
Molecular Weight (g/mol) | 130.143 |
MDL Number | MFCD00009199 |
InChI Key | XYIBRDXRRQCHLP-UHFFFAOYSA-N |
Synonym | ethyl acetoacetate, ethyl acetylacetate, ethyl 3-oxobutyrate, diacetic ether, butanoic acid, 3-oxo-, ethyl ester, ethyl acetylacetonate, acetoacetic acid, ethyl ester, 3-oxobutanoic acid ethyl ester, 3-oxo-butyric acid ethyl ester, active acetylacetate |
PubChem CID | 8868 |
ChEBI | CHEBI:4893 |
IUPAC Name | ethyl 3-oxobutanoate |
SMILES | CCOC(=O)CC(=O)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11458940
|
Thermo Scientific Alfa Aesar
A12544.30 |
250 g |
24.82 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11494224
|
Thermo Scientific Alfa Aesar
A12544.36 |
500 g |
29.65 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11404234
|
Thermo Scientific Alfa Aesar
A12544.0E |
2500 g |
80.10 €
2500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11414234
|
Thermo Scientific Alfa Aesar
A12544.0C |
10,000 g |
274.37 €
10000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ethyl acetoacetate is used as an intermediate in organic synthesis and as a co-promoter for unsaturated polyester resins. It is widely used in the production of dyes, inks, perfumes, plastics and flavoring agents. It is an important starting material for the syntheses of alpha-substituted acetoacetic esters and cyclic compounds like pyrazole, pyrimidine and coumarin derivatives. It acts as an intermediate in the synthesis of vitamins and pharmaceuticals. It finds application as a formaldehyde scavenger.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEthyl acetoacetate is used as an intermediate in organic synthesis and as a co-promoter for unsaturated polyester resins. It is widely used in the production of dyes, inks, perfumes, plastics and flavoring agents. It is an important starting material for the syntheses of alpha-substituted acetoacetic esters and cyclic compounds like pyrazole, pyrimidine and coumarin derivatives. It acts as an intermediate in the synthesis of vitamins and pharmaceuticals. It finds application as a formaldehyde scavenger.
Solubility
Miscible with water, benzene, chloroform, ether and acetone.
Notes
Incompatible with acids, bases, oxidizing agents, alkali metals and reducing agents.
Chemical Identifiers
141-97-9 | |
130.143 | |
XYIBRDXRRQCHLP-UHFFFAOYSA-N | |
8868 | |
ethyl 3-oxobutanoate |
C6H10O3 | |
MFCD00009199 | |
ethyl acetoacetate, ethyl acetylacetate, ethyl 3-oxobutyrate, diacetic ether, butanoic acid, 3-oxo-, ethyl ester, ethyl acetylacetonate, acetoacetic acid, ethyl ester, 3-oxobutanoic acid ethyl ester, 3-oxo-butyric acid ethyl ester, active acetylacetate | |
CHEBI:4893 | |
CCOC(=O)CC(=O)C |
Specifications
141-97-9 | |
1.025 | |
84°C (183°F) | |
Fruit-like | |
1.419 | |
385838 | |
ethyl acetoacetate, ethyl acetylacetate, ethyl 3-oxobutyrate, diacetic ether, butanoic acid, 3-oxo-, ethyl ester, ethyl acetylacetonate, acetoacetic acid, ethyl ester, 3-oxobutanoic acid ethyl ester, 3-oxo-butyric acid ethyl ester, active acetylacetate | |
XYIBRDXRRQCHLP-UHFFFAOYSA-N | |
ethyl 3-oxobutanoate | |
8868 | |
130.14 | |
Liquid |
-43°C | |
180°C to 181°C | |
≥99% | |
C6H10O3 | |
MFCD00009199 | |
14,3758 | |
Miscible with water,benzene,chloroform,ether and acetone. | |
CCOC(=O)CC(=O)C | |
130.143 | |
CHEBI:4893 | |
≥99% | |
Ethyl acetoacetate |
Safety and Handling
GHS H Statement
H319-H227
Causes serious eye irritation.
Combustible liquid.
P210-P280-P370+P378q-P501c
H227
missing translation for 'einecsNumber' : 205-516-1
missing translation for 'rtecsNumber' : AK5250000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.