missing translation for 'onlineSavingsMsg'
Learn More

Ethyl acrylate, 99.5%, stabilized

Product Code. 10132910 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 mL
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
2.5 L
25mL
This item is not returnable. View return policy

Product Code. 10132910

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 140-88-5 | C5H8O2 | 100.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 140-88-5, 150-76-5
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00009188
InChI Key JIGUQPWFLRLWPJ-UHFFFAOYSA-N
Synonym ethyl acrylate, acrylic acid ethyl ester, ethyl propenoate, 2-propenoic acid, ethyl ester, ethyl 2-propenoate, ethylacrylaat, ethylakrylat, etil acrilato, acrylic acid, ethyl ester, aethylacrylat
PubChem CID 8821
ChEBI CHEBI:82327
IUPAC Name ethyl prop-2-enoate
SMILES CCOC(=O)C=C

Specifications

Melting Point -71°C
Density 0.9200g/mL
Boiling Point 99°C
Flash Point 16°C
Infrared Spectrum Authentic
Assay Percent Range 99.40% min. (GC)
Packaging Glass bottle
Linear Formula H2C=CHCO2C2H5
Refractive Index 1.4050 to 1.407
Quantity 25 mL
Beilstein 02, 399
Fieser 06,251; 13,272
Merck Index 15, 3813
Specific Gravity 0.92
Solubility Information Solubility in water: 1.5% (25°C). Other solubilities: freely miscible with most organic solvents
Stabilizer 10 to 20ppm MEHQ
Viscosity 0.58 mPa.s (20°C)
Formula Weight 100.12
Percent Purity 99.50%
Physical Form Liquid
Chemical Name or Material Ethyl acrylate, Stabilized
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ethyl acrylate, stabilized
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Acute toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H317-May cause an allergic skin reaction.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H331-Toxic if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.