Learn More
Ethyl acrylate, 99%, stab. with ca 20ppm 4-methoxyphenol, Thermo Scientific Chemicals
Description
Ethyl acrylate is used in the syntheses of various pharmaceutical intermediates, textiles and non-woven fibers. It is widely used in the production of polymers including resins, plastics, rubber and denture material. It is actively involved in the preparation of dimethylaminoethylacrylate (DMAEA) which is used as a monomer to make flocculants for water treatment. It plays a vital role in the manufacture of water-based latex paints and adhesives, textile and paper coatings and leather finish resins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Ethyl acrylate is used in the syntheses of various pharmaceutical intermediates, textiles and non-woven fibers. It is widely used in the production of polymers including resins, plastics, rubber and denture material. It is actively involved in the preparation of dimethylaminoethylacrylate (DMAEA) which is used as a monomer to make flocculants for water treatment. It plays a vital role in the manufacture of water-based latex paints and adhesives, textile and paper coatings and leather finish resins.
Solubility
Miscible with ethanol, ethyl ether and chloroform. Silghtly miscible with water and dimethyl sulfoxide.
Notes
Store in cool place. Incompatible with strong oxidizing agents, strong acids, amines, peroxides, chlorosulfonic acid, moisture and strong alkalies.

Specifications
Specifications
Melting Point | -71°C |
Density | 0.924 |
Boiling Point | 99°C to 100°C |
Flash Point | −1°C (30°F) |
Odor | Pungent |
Linear Formula | CH2=CHCO2CH2CH3 |
Refractive Index | 1.406 |
Quantity | 100 mL |
UN Number | UN1917 |
Beilstein | 773866 |
Show More |
Safety and Handling


- Ethyl acrylate
- Danger
- Acute toxicity Category 3
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H335-May cause respiratory irritation.
- H412-Harmful to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- MIXTURE LIST-Contains : Ethyl acrylate
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.