Learn More
Ethyl alpha,4-dibromophenylacetate, 97+%, Thermo Scientific Chemicals
143.00 € - 484.00 €
Chemical Identifiers
CAS | 77143-76-1 |
---|---|
Molecular Formula | C10H10Br2O2 |
Molecular Weight (g/mol) | 321.996 |
MDL Number | MFCD09953497 |
InChI Key | PPRAWWBOUOBAON-UHFFFAOYSA-N |
Synonym | ethyl 2-bromo-2-4-bromophenyl acetate, ethyl 2-bromo-4-bromophenyl acetate, ethyl bromo 4-bromophenyl acetate, alpha,4-dibromophenylacetic acid ethyl ester, ethyl alpha,4-dibromophenylacetate, ethyl2-bromo-2-4-bromophenyl acetate, ethyl 2-bromanyl-2-4-bromophenyl ethanoate, benzeneacetic acid, ?,4-dibromo-, ethyl ester, benzeneacetic acid, a,4-dibromo-, ethyl ester |
PubChem CID | 12754876 |
IUPAC Name | ethyl 2-bromo-2-(4-bromophenyl)acetate |
SMILES | CCOC(=O)C(C1=CC=C(C=C1)Br)Br |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11397579
|
Thermo Scientific Alfa Aesar
H28093.06 |
5 g |
143.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11307589
|
Thermo Scientific Alfa Aesar
H28093.14 |
25 g |
484.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is employed as intermediate for pharmaceutical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is employed as intermediate for pharmaceutical.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Chemical Identifiers
77143-76-1 | |
321.996 | |
PPRAWWBOUOBAON-UHFFFAOYSA-N | |
12754876 | |
CCOC(=O)C(C1=CC=C(C=C1)Br)Br |
C10H10Br2O2 | |
MFCD09953497 | |
ethyl 2-bromo-2-4-bromophenyl acetate, ethyl 2-bromo-4-bromophenyl acetate, ethyl bromo 4-bromophenyl acetate, alpha,4-dibromophenylacetic acid ethyl ester, ethyl alpha,4-dibromophenylacetate, ethyl2-bromo-2-4-bromophenyl acetate, ethyl 2-bromanyl-2-4-bromophenyl ethanoate, benzeneacetic acid, ?,4-dibromo-, ethyl ester, benzeneacetic acid, a,4-dibromo-, ethyl ester | |
ethyl 2-bromo-2-(4-bromophenyl)acetate |
Specifications
77143-76-1 | |
140°C (5 mmHg) | |
C10H10Br2O2 | |
MFCD09953497 | |
ethyl 2-bromo-2-4-bromophenyl acetate, ethyl 2-bromo-4-bromophenyl acetate, ethyl bromo 4-bromophenyl acetate, alpha,4-dibromophenylacetic acid ethyl ester, ethyl alpha,4-dibromophenylacetate, ethyl2-bromo-2-4-bromophenyl acetate, ethyl 2-bromanyl-2-4-bromophenyl ethanoate, benzeneacetic acid, ?,4-dibromo-, ethyl ester, benzeneacetic acid, a,4-dibromo-, ethyl ester | |
PPRAWWBOUOBAON-UHFFFAOYSA-N | |
ethyl 2-bromo-2-(4-bromophenyl)acetate | |
12754876 | |
≥97% |
1.642 | |
>110°C (230°F) | |
1.571 | |
UN3265 | |
Slightly soluble in water. | |
CCOC(=O)C(C1=CC=C(C=C1)Br)Br | |
321.996 | |
321.99 | |
Ethyl alpha,4-dibromophenylacetate |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only