missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl benzoate, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
2500 g
Description
Ethyl benzoate is used as a perfume scent. It acts as a food flavoring agent. It is an active component of artificial fruit flavors. Further, it is used in cosmetics and personal care products as fragrance ingredients and preservatives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ethyl benzoate is used as a perfume scent. It acts as a food flavoring agent. It is an active component of artificial fruit flavors. Further, it is used in cosmetics and personal care products as fragrance ingredients and preservatives.
Solubility
Miscible with methanol and diethyl ether. Immiscible with water.
Notes
Incompatible with strong oxidizing agents and strong bases.
Ethyl benzoate is used as a perfume scent. It acts as a food flavoring agent. It is an active component of artificial fruit flavors. Further, it is used in cosmetics and personal care products as fragrance ingredients and preservatives.
Solubility
Miscible with methanol and diethyl ether. Immiscible with water.
Notes
Incompatible with strong oxidizing agents and strong bases.
Specifications
Specifications
Melting Point | -34°C |
Density | 1.049 |
Boiling Point | 212°C to 214°C |
Flash Point | 84°C (183°F) |
Refractive Index | 1.505 |
Quantity | 500 g |
Beilstein | 1908172 |
Merck Index | 14,3766 |
Solubility Information | Miscible with methanol and diethyl ether. Immiscible with water. |
Formula Weight | 150.18 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'