missing translation for 'onlineSavingsMsg'
Learn More

Ethyl chloroacetate, 99%

Product Code. 11472997
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
This item is not returnable. View return policy

Product Code. 11472997

Brand: Thermo Scientific Alfa Aesar A15554.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 105-39-5 | C4H7ClO2 | 122.548 g/mol

Ethyl chloroacetate is a reagent used in the preparation of 5 member heterocycles. It is used as pharmaceutical and organic intermediate. It is used as a solvent for organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl chloroacetate is a reagent used in the preparation of 5 member heterocycles. It is used as pharmaceutical and organic intermediate. It is used as a solvent for organic synthesis.

Solubility
Soluble in water 20 g/L (20°C).

Notes
Incompatible with acids, bases, oxidizing agents, reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105-39-5
Molecular Formula C4H7ClO2
Molecular Weight (g/mol) 122.548
MDL Number MFCD00000932
InChI Key VEUUMBGHMNQHGO-UHFFFAOYSA-N
Synonym ethyl chloroacetate, ethyl chloracetate, ethyl monochloroacetate, acetic acid, chloro-, ethyl ester, chloroacetic acid ethyl ester, ethyl monochloracetate, ethyl chloroethanoate, ethyl alpha-chloroacetate, 2-chloroacetic acid ethyl ester, unii-6h07teh4tf
PubChem CID 7751
IUPAC Name ethyl 2-chloroacetate
SMILES CCOC(=O)CCl

Specifications

Melting Point -26°C
Density 1.159
Boiling Point 142°C to 144°C
Flash Point 54°C (149°F)
Odor Pungent
Linear Formula ClCH2CO2CH2CH3
Refractive Index 1.421
Quantity 500 g
UN Number UN1181
Beilstein 506455
Merck Index 14,3783
Solubility Information Soluble in water 20g/L (20°C).
Formula Weight 122.55
Percent Purity 99%
Chemical Name or Material Ethyl chloroacetate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.