Learn More
Ethyl chloroacetate, 99%, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | -26.0°C |
Color | Colorless |
Density | 1.1500g/mL |
Boiling Point | 142.0°C to 145.0°C |
Flash Point | 53°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 99% |
Packaging | Glass bottle |
Molecular Formula | C4H7ClO2 |
Linear Formula | ClCH2CO2C2H5 |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0469331 | Certificate of Analysis | 31/01/2025 | 118220100,118222500,118220025,118220010 |
A0406904 | Certificate of Analysis | 12/12/2022 | 118220100,118222500,118220025,118220010 |
A0447307 | Certificate of Analysis | 07/11/2022 | 118220100,118222500,118220025,118220010 |
A0436858 | Certificate of Analysis | 20/07/2022 | 118220100,118222500,118220025,118220010 |
A0442719 | Certificate of Analysis | 10/06/2022 | 118220100,118222500,118220025,118220010 |
Safety and Handling




- Ethyl chloroacetate
- Danger
- Acute toxicity Category 3
- ACUTE AQUATIC Acute 1
- Flammable liquid Category 3
- Acute toxicity Category 2
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
- H400-Very toxic to aquatic life.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H226-Flammable liquid and vapour.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.